Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166456.1 | NP_001159928.1 | 3 | 468 | ||
NM_001166458.1 | NP_001159930.1 | 3 | 675 | |||
NM_134086.4 | NP_598847.2 | 3 | 1161 | |||
GenBank mRNA | AF184240.2 | - | 2 | 253 | ||
AK029189.1 | - | 3 | 338 | |||
AK034130.1 | - | 3 | 1163 | |||
AK035098.1 | - | 2 | 180 | |||
AK047459.1 | - | 3 | 692 | |||
AK049173.1 | - | 3 | 375 | |||
AK049294.1 | - | 3 | 441 | |||
AK050914.1 | - | 3 | 376 | |||
AK081724.1 | - | 3 | 677 | |||
AK082683.1 | - | 3 | 376 | |||
AK162612.1 | - | 3 | 526 | |||
AK165188.1 | - | 3 | 728 | |||
AK199869.1 | - | 2 | 92 | |||
BC030378.1 | - | 3 | 377 | 5375319 | ||
BC066815.1 | - | 3 | 338 |