Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134138.1 | NP_598899.1 | 5 | 638 | ||
GenBank mRNA | AB031387.1 | - | 5 | 638 | ||
AK077569.1 | - | 5 | 625 | |||
AK167840.1 | - | 5 | 879 |