Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Desi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134095.2 | NP_598856.1 | 6 | 791 | ||
GenBank mRNA | AK005706.1 | - | 6 | 782 | ||
AK005963.1 | - | 6 | 747 | |||
AK016347.1 | - | 6 | 697 | |||
AK131703.1 | - | 6 | 745 | |||
AK146155.1 | - | 6 | 815 | |||
AK170731.1 | - | 7 | 1060 | |||
BC022097.1 | - | 5 | 746 | |||
BC056972.1 | - | 6 | 791 | 6811343 | ||
BC071205.1 | - | 6 | 683 | 6515068 | ||
BC131979.1 | - | 6 | 687 | 40130256 | ||
BC132642.1 | - | 6 | 687 | 40130919 | ||
BC145235.1 | - | 5 | 617 |