Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290536.1 | NP_001277465.1 | 2 | 954 | ||
NM_001290537.1 | NP_001277466.1 | 3 | 574 | |||
NM_138604.3 | NP_613070.2 | 2 | 954 | |||
GenBank mRNA | AF229642.1 | - | 2 | 95 | ||
AK010750.1 | - | 3 | 572 | |||
AK149777.1 | - | 3 | 575 | |||
AK155131.1 | - | 2 | 930 | |||
AK167226.1 | - | 2 | 180 | |||
AK169921.1 | - | 3 | 540 | |||
AK171874.1 | - | 2 | 695 | |||
AK212697.1 | - | 1 | 47 | |||
BC016529.1 | - | 2 | 954 | 3594583 | ||
BC051111.1 | - | 2 | 693 | 5253679 |