Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Havcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134250.2 | NP_599011.2 | 7 | 1575 | ||
GenBank mRNA | AF450241.1 | - | 7 | 1571 | ||
AK148474.1 | - | 7 | 1576 | |||
AK170775.1 | - | 8 | 1396 |