Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r3f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290574.1 | NP_001277503.1 | 4 | 2561 | ||
NM_138605.3 | NP_613071.3 | 4 | 2558 | |||
GenBank mRNA | AF229644.1 | - | 4 | 2397 | ||
AK147873.1 | - | 1158 | ||||
BC059275.1 | - | 4 | 2495 | |||
BC138138.1 | - | 4 | 2390 | 8861157 |