Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam50a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138607.3 | NP_613073.2 | 6, 7 | 763 | ||
GenBank mRNA | AK008402.1 | - | 6, 7 | 764 | ||
BC026128.1 | - | 5, 6 | 1136 | |||
BC125297.1 | - | 5, 6 | 629 | 40129812 | ||
Y18505.1 | - | 6, 7 | 728 |