Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138646.3 | NP_619587.3 | 7, 8 | 1136 | ||
GenBank mRNA | AK081489.1 | - | 7, 8 | 1096 | ||
AK129418.1 | - | 6, 7 | 1023 | |||
AK147954.1 | - | 6, 7 | 1579 | |||
AK171871.1 | - | 7, 8 | 1105 | |||
AY043415.1 | - | 6, 7 | 656 | |||
BC063055.1 | - | 6, 7 | 828 | 5701206 |