Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139219.2 | NP_631965.1 | 2 | 160 | ||
GenBank mRNA | AJ437647.1 | - | 2 | 147 | ||
BC109177.2 | - | 2 | 143 | 40049218 | ||
BM936051.1 | - | 2 | 189 |