Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138669.1 | NP_619610.1 | 3, 4 | 364 | ||
GenBank mRNA | AK075920.1 | - | 3, 4 | 340 | ||
AK150277.1 | - | 3, 4 | 380 | |||
AK151786.1 | - | 3, 4 | 346 | |||
AK152824.1 | - | 3, 4 | 380 | |||
AK153359.1 | - | 3, 4 | 346 | |||
AK157855.1 | - | 3, 4 | 340 | |||
AK167107.1 | - | 3, 4 | 402 | |||
AK168319.1 | - | 3, 4 | 344 | |||
AK169350.1 | - | 3, 4 | 344 | |||
AK169815.1 | - | 1770 | ||||
BC008132.1 | - | 3, 4 | 364 | 3498954 | ||
BC012862.1 | - | 3, 4 | 352 | 3585696 |
Gene Symbol: Gm5580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC158083.1 | - | 1 | 340 | 9088173 |
Gene Symbol: Gm8994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142734.1 | NP_001136206.1 | 2 | 449 | ||
GenBank mRNA | AK139713.1 | - | 2 | 450 |