Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab40c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139154.2 | NP_631893.1 | 4 | 338 | ||
GenBank mRNA | AB232639.1 | - | 3 | 232 | ||
AF422144.1 | - | 3 | 291 | |||
AK133766.1 | - | 3 | 114 | |||
BC027008.1 | - | 4 | 326 | 5364768 | ||
BC037461.1 | - | 4 | 459 |