Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139200.4 | NP_631939.1 | 2, 3 | 299 | ||
GenBank mRNA | AF192525.1 | - | 2, 3 | 219 | ||
AK089364.1 | - | 2, 3 | 240 | |||
AK152784.1 | - | 2, 3 | 281 | |||
AK157859.1 | - | 2, 3 | 300 | |||
AK164790.1 | - | 3, 4 | 305 | |||
AK166254.1 | - | 2, 3 | 300 | |||
AK168343.1 | - | 2, 3 | 299 | |||
AK169575.1 | - | 2, 3 | 285 | |||
AK170544.1 | - | 2, 3 | 300 | |||
AK170796.1 | - | 2, 3 | 300 | |||
AK171467.1 | - | 2, 3 | 300 | |||
AK172071.1 | - | 2, 3 | 356 | |||
AK172380.1 | - | 2, 3 | 300 | |||
BC007144.1 | - | 2, 3 | 288 | 3487179 |