Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139272.2 | NP_644678.2 | 2 | 319 | ||
GenBank mRNA | AF348968.1 | - | 2 | 187 | ||
AK046567.1 | - | 2 | 277 | |||
AK145122.1 | - | 2 | 197 | |||
AK149609.1 | - | 2 | 245 | |||
AK151536.1 | - | 2 | 110 | |||
BC007172.1 | - | 1 | 54 | |||
BC053063.1 | - | 1 | 162 | 5702572 | ||
BC059818.1 | - | 2 | 264 | 5697789 | ||
BQ770019.1 | - | 2 | 319 |