Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000624.5 | NP_000615.3 | 6 | 1561 | ||
GenBank mRNA | AK096131.1 | - | 6 | 1266 | ||
AK293192.1 | - | 7 | 1256 | |||
AK298279.1 | - | 6 | 1167 | |||
AL080185.1 | - | 4 | 1245 | |||
BC008915.2 | - | 6 | 1456 | 3618167 |