Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK222954.1 | - | 5 | 654 | ||
AK223210.1 | - | 5 | 659 | |||
AK313583.1 | - | 5 | 651 | |||
AX775815.1 | - | 5 | 650 | |||
AX775817.1 | - | 5 | 647 | |||
BC002975.1 | - | 5 | 634 | 3542354 | ||
BC032651.1 | - | 5 | 617 | 5504755 | ||
M80461.1 | - | 5 | 597 | |||
M89957.1 | - | 5 | 675 | |||
S52229.1 | - | 5 | 581 | |||
S79249.1 | - | 4 | 269 | |||
X83539.1 | - | 4 | 269 |