Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144789.2 | NP_659038.2 | 21 | 2786 | ||
GenBank mRNA | AK144820.1 | - | 21 | 2691 | ||
AY063457.1 | - | 2 | 344 | |||
BC046433.1 | - | 2 | 345 | |||
BC072597.1 | - | 19 | 2473 |