Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159415.1 | NP_001152887.1 | 12 | 1856 | ||
NM_144511.2 | NP_653094.2 | 13 | 2006 | |||
GenBank mRNA | BC010812.1 | - | 12 | 1846 | ||
BC019147.1 | - | 13 | 2002 | 5052937 | ||
BC019926.1 | - | 13 | 1997 |