Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252478.1 | NP_001239407.1 | 14 | 1301 | ||
NM_144798.6 | NP_659047.2 | 14 | 1286 | |||
GenBank mRNA | AF233346.1 | - | 13 | 1238 | ||
AF395840.1 | - | 13 | 1228 | |||
AK082907.1 | - | 14 | 1263 | |||
AK150342.1 | - | 14 | 1262 | |||
AK151120.1 | - | 14 | 1262 | |||
AK160903.1 | - | 14 | 1290 | |||
BC005753.1 | - | 6 | 749 | |||
BC066162.1 | - | 14 | 1253 | 30471027 | ||
BC068169.1 | - | 14 | 1275 | 30542277 |