Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000666.2 | NP_000657.1 | 4, 5 | 596 | ||
NM_001198895.1 | NP_001185824.1 | 4, 5 | 591 | |||
NM_001198896.1 | NP_001185825.1 | 4, 5 | 596 | |||
NM_001198897.1 | NP_001185826.1 | 4, 5 | 596 | |||
NM_001198898.1 | NP_001185827.1 | 3, 4 | 491 | |||
GenBank mRNA | AK298274.1 | - | 4, 5 | 345 | ||
BC000545.2 | - | 4, 5 | 340 | 3162222 | ||
BC003023.1 | - | 4, 5 | 333 | 2821665 | ||
BC014112.2 | - | 4, 5 | 316 | 4543442 | ||
CD014023.1 | - | 4, 5 | 437 | |||
CD014026.1 | - | 4, 5 | 437 | |||
CD014027.1 | - | 3, 4 | 332 | |||
D14524.1 | - | 4, 5 | 330 | |||
D16307.1 | - | 4, 5 | 328 | |||
EU794586.1 | - | 4, 5 | 316 | |||
KU177890.1 | - | 3, 4 | 272 | |||
L07548.1 | - | 4, 5 | 333 |
Gene Symbol: ABHD14A-ACY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316331.1 | NP_001303260.1 | 6, 7 | 797 | ||
GenBank mRNA | AK298086.1 | - | 6, 7 | 773 |