Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164566.1 | NP_001158038.1 | 5 | 352 | ||
NM_144882.4 | NP_659131.2 | 7 | 538 | |||
GenBank mRNA | AK031266.1 | - | 7 | 493 | ||
AK032169.1 | - | 7 | 473 | |||
BC014764.1 | - | 5 | 352 | 4217648 | ||
BC021879.1 | - | 7 | 474 | 4216816 | ||
BY069430.1 | - | 5 | 308 | |||
BY739337.1 | - | 5 | 315 | |||
EF410080.1 | - | 578 |