Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144811.3 | NP_659060.1 | 7 | 1114 | ||
GenBank mRNA | AK045714.1 | - | 6 | 1614 | ||
AY453793.1 | - | 7 | 1101 | |||
BC021398.1 | - | 7 | 1130 | 5066655 | ||
BC030919.1 | - | 2 | 1207 | |||
BC042657.1 | - | 4 | 4284 |