Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144818.3 | NP_659067.2 | 9 | 1142 | ||
GenBank mRNA | AK028919.1 | - | 9 | 1127 | ||
AK080433.1 | - | 9 | 1131 | |||
BC021499.1 | - | 9 | 1116 | 5346298 | ||
BC025060.1 | - | 9 | 1067 | 3673890 |