Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncaph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144818.3 | NP_659067.2 | 9 | 1187 | ||
GenBank mRNA | AK028919.1 | - | 9 | 1172 | ||
AK042706.1 | - | 9 | 1104 | |||
AK080433.1 | - | 9 | 1176 | |||
AK129047.1 | - | 6 | 740 | |||
AK187987.1 | - | 2 | 160 | |||
BC021499.1 | - | 9 | 1161 | 5346298 | ||
BC025060.1 | - | 9 | 1112 | 3673890 |