Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144819.2 | NP_659068.1 | 4 | 1686 | ||
GenBank mRNA | AK028192.1 | - | 4 | 1663 | ||
AK136203.1 | - | 4 | 1853 | |||
AK165371.1 | - | 4 | 1686 | |||
BC021492.1 | - | 4 | 1630 | 5345674 |