Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmym5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253752.1 | NP_001240681.1 | 8 | 1923 | ||
NM_001253753.1 | NP_001240682.1 | 8 | 1993 | |||
NM_144842.3 | NP_659091.3 | 8 | 1987 | |||
GenBank mRNA | AK036513.1 | - | 7 | 1832 | ||
AK156501.1 | - | 8 | 1996 | |||
AK165292.1 | - | 1 | 648 | |||
BC027380.1 | - | 6 | 1336 |