Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144868.3 | NP_659117.2 | 35 | 7183 | ||
GenBank mRNA | AF237953.1 | - | 33 | 5200 | ||
AK144114.1 | - | 11 | 2443 | |||
AK220471.1 | - | 13 | 3081 | |||
BC038056.1 | - | 30 | 5291 | |||
BC054751.1 | - | 35 | 6462 | |||
BC055289.1 | - | 36 | 6986 | |||
BC096538.1 | - | 35 | 7183 |