Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144887.4 | NP_659136.1 | 12 | 3520 | ||
GenBank mRNA | AK028737.1 | - | 9 | 2086 | ||
AK075641.1 | - | 2 | 866 | |||
AK082513.1 | - | 12 | 3445 | |||
AK145086.1 | - | 12 | 3448 | |||
AK173251.1 | - | 12 | 3475 | |||
AY894891.1 | - | 12 | 3512 | |||
BC020051.1 | - | 11 | 2490 | 3987696 | ||
BC027047.1 | - | 8 | 1754 | |||
BC065155.1 | - | 12 | 3390 | 5706087 |