Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144890.2 | NP_659139.2 | 9, 10 | 1086 | ||
GenBank mRNA | AK142982.1 | - | 9, 10 | 1087 | ||
BC018465.1 | - | 9, 10 | 1048 | 3156626 |