Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144898.2 | NP_659147.2 | 7 | 676 | ||
GenBank mRNA | AK134001.1 | - | 133 | |||
AK139708.1 | - | 655 | ||||
AK163280.1 | - | 7 | 536 | |||
AK166834.1 | - | 1 | 2108 | |||
AK167586.1 | - | 7 | 624 | |||
AK170160.1 | - | 7 | 659 | |||
BC008103.1 | - | 7 | 626 | 3966815 |