Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144919.2 | NP_659168.1 | 2 | 171 | ||
GenBank mRNA | AK045994.1 | - | 2 | 183 | ||
AK177072.1 | - | 1 | 74 | |||
AK185494.1 | - | 1 | 74 | |||
AK217618.1 | - | 1 | 19 | |||
BC016208.1 | - | 2 | 171 | 4913322 |