Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145148.2 | NP_660130.2 | 4 | 727 | ||
GenBank mRNA | AB042027.1 | - | 4 | 650 | ||
AF327856.1 | - | 3 | 350 | |||
AF327857.1 | - | 2 | 137 | |||
AK051779.1 | - | 4 | 599 | |||
AK129262.1 | - | 3 | 202 | |||
AK132280.1 | - | 4 | 591 | |||
AK143978.1 | - | 3 | 349 | |||
BC052390.1 | - | 4 | 596 | 5718942 |