Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6330416G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144905.3 | NP_659154.2 | 10 | 2028 | ||
GenBank mRNA | AK030466.1 | - | 10 | 2158 | ||
AK031016.1 | - | 10 | 2032 | |||
AK078066.1 | - | 1 | 698 | |||
AK138872.1 | - | 8 | 1845 | |||
AK144990.1 | - | 11 | 2205 | |||
AK198553.1 | - | 1 | 146 | |||
AK205279.1 | - | 1 | 146 | |||
AK207062.1 | - | 1 | 147 | |||
AK209541.1 | - | 1 | 146 | |||
AK213527.1 | - | 1 | 146 | |||
AK214000.1 | - | 1 | 146 | |||
AK215224.1 | - | 1 | 146 | |||
BC022599.1 | - | 8 | 1882 | 4159235 | ||
BC023748.1 | - | 10 | 2200 | 5353894 |