Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mib2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256107.1 | NP_001243036.1 | 19 | 3437 | ||
NM_001256108.2 | NP_001243037.2 | 19 | 3257 | |||
NM_145124.3 | NP_660106.2 | 18 | 3341 | |||
GenBank mRNA | AB072336.1 | - | 18 | 2906 | ||
AK008671.1 | - | 5 | 1203 | |||
AK084791.1 | - | 10 | 1842 | |||
AK147940.1 | - | 19 | 3203 | |||
AK159611.1 | - | 19 | 3154 | |||
AY974090.1 | - | 17 | 2638 | |||
BC058086.1 | - | 16 | 2925 | |||
BC141133.1 | - | 16 | 2730 | 9055745 | ||
BC145441.1 | - | 17 | 2826 |