Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145126.2 | NP_660108.2 | 8 | 897 | ||
GenBank mRNA | AK131914.1 | - | 8 | 898 | ||
AY049765.2 | - | 8 | 882 | |||
AY065557.1 | - | 8 | 882 | |||
BC130015.1 | - | 8 | 886 | 40140436 |