Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145130.2 | NP_660112.1 | 13 | 1803 | ||
GenBank mRNA | AK083687.1 | - | 11 | 1359 | ||
AK098090.1 | - | 13 | 1774 | |||
AK154062.1 | - | 13 | 1797 | |||
AK165319.1 | - | 13 | 1782 | |||
AK169452.1 | - | 12 | 1920 | |||
AY028317.1 | - | 13 | 1780 | |||
BC006753.1 | - | 13 | 1731 | 3709076 |