Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145400.3 | NP_663375.3 | 4 | 529 | ||
GenBank mRNA | AK028721.1 | - | 4 | 506 | ||
AK028850.1 | - | 4 | 544 | |||
AK172893.1 | - | 4 | 489 | |||
BC006649.1 | - | 5 | 668 | 3498834 | ||
BC021406.1 | - | 4 | 535 | |||
BC025043.1 | - | 4 | 498 |