Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkag2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170556.1 | NP_001164027.1 | 1, 2 | 191 | ||
NM_145401.2 | NP_663376.2 | 5, 6 | 1273 | |||
GenBank mRNA | AK010703.1 | - | 1, 2 | 133 | ||
AK154296.1 | - | 5, 6 | 1274 | |||
AK165834.1 | - | 1, 2 | 133 | |||
AY348864.1 | - | 1, 2 | 124 | |||
BC015283.1 | - | 5, 6 | 1061 | 4238045 | ||
BQ572978.1 | - | 1, 2 | 124 |