Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145415.2 | NP_663390.2 | 3 | 418 | ||
GenBank mRNA | AK052355.1 | - | 3 | 419 | ||
AK088756.1 | - | 3 | 431 | |||
AK147251.1 | - | 3 | 412 | |||
BC019218.1 | - | 4 | 378 | 5065857 |