Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acnat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145368.2 | NP_663343.2 | 5 | 1429 | ||
GenBank mRNA | AK050308.1 | - | 5 | 1429 | ||
AK050357.1 | - | 6 | 1426 | |||
BC010829.1 | - | 3 | 1273 | 4220620 |