Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC022126.1 | - | 4 | 500 | 5097371 | |
BC033482.1 | - | 2 | 254 | |||
BC145203.1 | - | 2 | 641 |