Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145476.2 | NP_663451.2 | 6 | 876 | ||
GenBank mRNA | AK039627.1 | - | 6 | 889 | ||
AK146872.1 | - | 6 | 877 | |||
AK154226.1 | - | 6 | 898 | |||
AK154602.1 | - | 6 | 919 | |||
AK155459.1 | - | 6 | 890 | |||
AK155770.1 | - | 6 | 919 | |||
AK178601.1 | - | 2 | 101 | |||
AK194823.1 | - | 2 | 101 | |||
AK194954.1 | - | 285 | ||||
AK208524.1 | - | 2 | 101 | |||
AK208550.1 | - | 2 | 101 | |||
AK210797.1 | - | 2 | 101 | |||
AK214703.1 | - | 2 | 101 | |||
AK216899.1 | - | 2 | 101 | |||
AK216948.1 | - | 2 | 101 | |||
AK217113.1 | - | 2 | 101 | |||
BC023106.1 | - | 3 | 271 | 4016550 | ||
BC066009.1 | - | 6 | 876 | 5708486 |