Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2j13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145548.4 | NP_663523.2 | 3, 4 | 599 | ||
GenBank mRNA | AK143903.1 | - | 3, 4 | 600 | ||
AK143981.1 | - | 3, 4 | 596 | |||
AK162358.1 | - | 603 | ||||
AK165405.1 | - | 3, 4 | 599 | |||
AK165469.1 | - | 3, 4 | 577 | |||
AK165473.1 | - | 3, 4 | 577 | |||
BC016446.1 | - | 3, 4 | 589 | 4235131 | ||
BC024451.1 | - | 2, 3 | 219 |