Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145479.4 | NP_663454.3 | 3 | 394 | ||
GenBank mRNA | AK028197.1 | - | 3 | 893 | ||
AK030669.1 | - | 3 | 339 | |||
AK086748.1 | - | 3 | 341 | |||
AK143514.1 | - | 3 | 339 | |||
AK204425.1 | - | 2 | 120 | |||
AK206875.1 | - | 2 | 120 | |||
AK214791.1 | - | 2 | 120 | |||
BC005800.1 | - | 3 | 372 | 2812589 | ||
CJ159851.1 | - | 3 | 386 |