Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145569.4 | NP_663544.1 | 1, 2 | 212 | ||
GenBank mRNA | AB070266.1 | - | 1, 2 | 176 | ||
AK034757.1 | - | 1, 2 | 213 | |||
AK052764.1 | - | 1, 2 | 211 | |||
AK079420.1 | - | 1, 2 | 218 | |||
AK133960.1 | - | 1, 2 | 211 | |||
AK144189.1 | - | 1, 2 | 215 | |||
AK145407.1 | - | 1, 2 | 211 | |||
AK150040.1 | - | 1, 2 | 211 | |||
AK151281.1 | - | 1, 2 | 211 | |||
AK160869.1 | - | 1, 2 | 211 | |||
AK161852.1 | - | 1, 2 | 211 | |||
AK164137.1 | - | 1, 2 | 211 | |||
AK166671.1 | - | 1, 2 | 211 | |||
AK168155.1 | - | 1, 2 | 211 | |||
AK169705.1 | - | 1, 2 | 211 | |||
AK190931.1 | - | 1, 2 | 41 | |||
AK200511.1 | - | 1, 2 | 117 | |||
AK206552.1 | - | 1, 2 | 41 | |||
AK206774.1 | - | 1, 2 | 41 | |||
AK213948.1 | - | 1, 2 | 41 | |||
AK216312.1 | - | 1, 2 | 41 | |||
AK219100.1 | - | 1, 2 | 41 | |||
BC003451.1 | - | 1, 2 | 152 | 2655444 | ||
BC058360.1 | - | 1, 2 | 193 | 6821781 |