Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145605.2 | NP_663580.2 | 2 | 253 | ||
GenBank mRNA | AK089954.1 | - | 2 | 216 | ||
AK161674.1 | - | 2 | 229 | |||
BC012312.1 | - | 2 | 253 | 2631632 | ||
BC023738.1 | - | 2 | 224 | 5352770 | ||
BC058359.1 | - | 2 | 224 | 6815650 | ||
BY086869.1 | - | 2 | 239 |