Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLVRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000712.3 | NP_000703.2 | 4, 5 | 345 | ||
NM_001253823.1 | NP_001240752.1 | 5, 6 | 439 | |||
XM_011515474.2 | XP_011513776.1 | 4, 5 | 347 | |||
XM_017012520.1 | XP_016868009.1 | 4, 5 | 354 | |||
GenBank mRNA | AK291862.1 | - | 5, 6 | 426 | ||
BC005902.1 | - | 4, 5 | 338 | 4282476 | ||
BC008456.1 | - | 4, 5 | 338 | 4249842 | ||
BM710258.1 | - | 4, 5 | 345 | |||
U34877.1 | - | 4, 5 | 318 | |||
X93086.1 | - | 4, 5 | 322 |