Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145705.3 | NP_663751.2 | 3 | 550 | ||
GenBank mRNA | AF214013.1 | - | 3 | 541 | ||
AF518764.1 | - | 3 | 515 | |||
AK011154.1 | - | 3 | 523 | |||
AK016456.1 | - | 3 | 483 | |||
AK028952.1 | - | 3 | 521 | |||
AK090324.1 | - | 3 | 517 | |||
AK144592.1 | - | 3 | 479 | |||
AK169480.1 | - | 1 | 65 | |||
AK186139.1 | - | 3 | 161 | |||
BC017142.1 | - | 1 | 738 | |||
BC030347.1 | - | 3 | 476 | 5003804 |