Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145743.2 | NP_665686.2 | 2 | 438 | ||
GenBank mRNA | AF397909.1 | - | 1 | 141 | ||
AF520417.1 | - | 2 | 287 | |||
AK044595.1 | - | 2 | 314 | |||
AK170996.1 | - | 2 | 289 | |||
BC023308.1 | - | 1 | 115 | |||
BC089595.1 | - | 2 | 288 | 30118411 |