Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257639.2 | XP_005257696.1 | 3 | 288 | ||
XM_011525183.2 | XP_011523485.1 | 3 | 277 | |||
XM_017025012.1 | XP_016880501.1 | 3 | 277 | |||
GenBank mRNA | AK289715.1 | - | 3 | 287 | ||
AK298710.1 | - | 3 | 287 | |||
BC057792.1 | - | 3 | 293 | 30331755 | ||
BC069649.1 | - | 3 | 240 | 7262441 | ||
BC074768.2 | - | 3 | 217 | 30915189 | ||
CR541766.1 | - | 3 | 194 | |||
DA113846.1 | - | 3 | 288 | |||
M83670.1 | - | 3 | 241 |