siRNA Details
Gene Symbol: Aida | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181732.4 | NP_859421.1 | 2 | 602 | ||
GenBank mRNA | AK076031.1 | - | 2 | 254 | ||
AK081494.1 | - | 2 | 603 | |||
AK143716.1 | - | 2 | 142 | |||
AK149788.1 | - | 2 | 354 | |||
AK150639.1 | - | 2 | 290 | |||
AK150702.1 | - | 2 | 351 | |||
AK151783.1 | - | 2 | 293 | |||
AK152961.1 | - | 2 | 293 | |||
AK153448.1 | - | 2 | 290 | |||
AK158103.1 | - | 2 | 236 | |||
AK159318.1 | - | 2 | 614 | |||
AK167520.1 | - | 2 | 319 | |||
AK172451.1 | - | 2 | 295 | |||
BC057183.1 | - | 2 | 257 | 5065689 | ||
BC066829.1 | - | 2 | 141 | |||
BC086763.1 | - | 2 | 307 | 5707830 |

More Information
Other Species Targeted

